Neohesperidin dihydrochalcone ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
NHDC, NCI-c60764, Neohesperidin DC, 1-(4-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-2,6-dihydroxyphenyl)-3-(3-hydroxy-4-methoxyphenyl)propan-1-one
N0675-25G
N0675-5G
TCIAN0675-25GEA
3400
NOK
TCIAN0675-25G
TCIAN0675-5G
Neohesperidin dihydrochalcone ≥98.0% (by titrimetric analysis)
Neohesperidin dihydrochalcone
Formel:
C₂₈H₃₆O₁₅ MW: 612,58 g/mol |
MDL nummer:
MFCD03840557 CAS-nummer: 20702-77-6 EINECS: 243-978-6 Merck Indeks: 14,06452 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on dried substance) |
Solubility in hot Water | almost transparency |
Drying loss | max. 11.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...