2,3-Norbornanedicarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Bicyclo[2.2.1]heptane-2,3-dicarboxylic Acid

N0753-25G N0753-5G
TCIAN0753-25GEA 1800 NOK
TCIAN0753-25G TCIAN0753-5G
2,3-Norbornanedicarboxylic acid ≥98.0% (by GC, titration analysis)
2,3-Norbornanedicarboxylic acid
Formel: C₉H₁₂O₄
MW: 184,19 g/mol
MDL nummer: MFCD00520577
CAS-nummer: 1724-08-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR