5-Norbornene-2-exo,3-exo-dimethanol ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
5-exo,6-exo-Bis(hydroxymethyl)-2-norbornene, Bicyclo[2.2.1]hept-5-ene-2-exo,3-exo-dimethanol
N0769-1G
N0769-5G
TCIAN0769-1GEA
930
NOK
TCIAN0769-1G
TCIAN0769-5G
5-Norbornene-2-exo,3-exo-dimethanol ≥97.0% (by GC)
5-Norbornene-2-exo,3-exo-dimethanol
Formel:
C₉H₁₄O₂ MW: 154,21 g/mol |
MDL nummer:
MFCD00799568 CAS-nummer: 699-95-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...