1,4,5,8-Naphthalenetetracarboxylic acid >60.0% (by NMR) (contains Monoanhydride)

Leverandør: TCI
Warning

Synonymer: Naphthalene-1,4,5,8-tetracarboxylic acid

N0770-25G N0770-250G
TCIAN0770-25GEA 890 NOK
TCIAN0770-25G TCIAN0770-250G
1,4,5,8-Naphthalenetetracarboxylic acid >60.0% (by NMR) (contains Monoanhydride)
1,4,5,8-Naphthalenetetracarboxylic acid
Formel: C₁₄H₈O₈
MW: 304,21 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00004012
CAS-nummer: 128-97-2
EINECS: 204-924-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (NMR) min. 60.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR