2-Nitroamino-2-imidazoline
Leverandør: TCI
|
Warning
|
Synonymer:
2-(Nitroimino)imidazolidine
N0858-25G
N0858-5G
TCIAN0858-25GEA
1875
NOK
TCIAN0858-25G
TCIAN0858-5G
2-Nitroamino-2-imidazoline
2-Nitroamino-2-imidazoline
Formel:
C₃H₆N₄O₂ MW: 130.11 g/mol Smeltepunkt: 222°C |
MDL nummer:
MFCD00047040 CAS-nummer: 5465-96-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Elemental analysis (Nitrogen) | 42.20 to 43.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...