Nitrofurantoin ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 1-[(5-Nitrofurfurylidene)amino]hydantoin

N0883-25G N0883-1G
TCIAN0883-25GEA 435 NOK
TCIAN0883-25G TCIAN0883-1G
Nitrofurantoin ≥98.0% (by HPLC, titration analysis)
Nitrofurantoin
Formel: C₈H₆N₄O₅
MW: 238.16 g/mol
Smeltepunkt: 268°C
MDL nummer: MFCD00003224
CAS-nummer: 67-20-9
EINECS: 200-646-5
Merck Indeks: 14,06599

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR