Nepafenac ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
2-Amino-3-benzoylbenzeneacetamide, 2-(2-Amino-3-benzoylphenyl)acetamide
N0932-200MG
N0932-1G
TCIAN0932-200MGEA
1150
NOK
TCIAN0932-200MG
TCIAN0932-1G
Nepafenac ≥98.0% (by HPLC, titration analysis)
Nepafenac
Formel:
C₁₅H₁₄N₂O₂ MW: 254,29 g/mol Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD08067732 CAS-nummer: 78281-72-8 Merck Indeks: 14,06469 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 182.0 to 186.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...