trans-β-Nitrostyrene ≥98.0% (by GC)

Leverandør: TCI
Warning

N0941-5G N0941-25G
TCIAN0941-5GEA 420 NOK
TCIAN0941-5G TCIAN0941-25G
trans-β-Nitrostyrene ≥98.0% (by GC)
trans-β-Nitrostyrene
Formel: C₈H₇NO₂
MW: 149,15 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00007402
CAS-nummer: 5153-67-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=750cd8a4136d0227012e63fb9148e02708056e7c&vid=750cd8a4136d0227012e63fb9148e02708056e7c&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR