3-(1-Naphthyl)acrylic acid ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
3-(Naphthalen-1-yl)acrylic acid
N0972-5G
N0972-25G
TCIAN0972-5GEA
3300
NOK
TCIAN0972-5G
TCIAN0972-25G
3-(1-Naphthyl)acrylic acid ≥95.0% (by GC)
3-(1-Naphthyl)acrylic acid
Formel:
C₁₃H₁₀O₂ MW: 198,22 g/mol |
MDL nummer:
MFCD00014317 CAS-nummer: 13026-12-5 EINECS: 235-887-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 93.0 % |
Melting point | 210.0 to 214.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...