9-(1-Naphthyl)anthracene ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
9-(Naphthalen-1-yl)anthracene
N0994-1G
N0994-5G
TCIAN0994-1GEA
905
NOK
TCIAN0994-1G
TCIAN0994-5G
9-(1-Naphthyl)anthracene ≥98.0% (by GC)
9-(1-Naphthyl)anthracene
Formel:
C₂₄H₁₆ MW: 304,39 g/mol |
MDL nummer:
MFCD19689379 CAS-nummer: 7424-70-6 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 163.0 to 166.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...