2-(2-Nitroanilino)ethanol ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: N-(2-Nitrophenyl)ethanolamine, 2-((2-Nitrophenyl)amino)ethanol, N-(2-Hydroxyethyl)-2-nitroaniline

N1000-5G N1000-25G
TCIAN1000-5GEA 635 NOK
TCIAN1000-5G TCIAN1000-25G
2-(2-Nitroanilino)ethanol ≥98.0% (by GC)
2-(2-Nitroanilino)ethanol
Formel: C₈H₁₀N₂O₃
MW: 182,18 g/mol
MDL nummer: MFCD00239475
CAS-nummer: 4926-55-0
EINECS: 225-555-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=251f1acdd479c52373b396f22258e3f76a088e72&vid=251f1acdd479c52373b396f22258e3f76a088e72&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR