O-(4-Nitrobenzoyl)hydroxylamine ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI

N1013-1G N1013-5G
TCIAN1013-1GEA 650 NOK
TCIAN1013-1G TCIAN1013-5G
O-(4-Nitrobenzoyl)hydroxylamine ≥97.0% (by HPLC, total nitrogen)
O-(4-Nitrobenzoyl)hydroxylamine
Formel: C₇H₆N₂O₄
MW: 182,14 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD11976095
CAS-nummer: 35657-36-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR