Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)

Leverandør: TCI
Danger

Synonymer: [NH2Me2][{RuCl((S)-tolbinap)}2(mu-Cl)3]

N1018-200MG N1018-1G
TCIAN1018-200MGEA 420 NOK
TCIAN1018-200MG TCIAN1018-1G
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formel: C₉₈H₈₈Cl₅NP₄Ru₂
MW: 1783,07 g/mol
MDL nummer: MFCD09753034
CAS-nummer: 309735-86-2

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR