Netilmicin sulphate ≥98.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: (2R,3R,4R,5R)-2-(((1S,2S,3R,4S,6R)-4-Amino-3-(((2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl)oxy)-6-(ethylamino)-2-hydroxycyclohexyl)oxy)-5-methyl-4-(methylamino)tetrahydro-2H-pyran-3,5-diol sulphate(2:5)

N1108-25MG N1108-100MG
TCIAN1108-25MGEA 875 NOK
TCIAN1108-25MG TCIAN1108-100MG
Netilmicin sulphate ≥98.0% (by HPLC)
Netilmicin sulphate
Formel: C₂₁H₄₁N₅O₇·⁵/₂H₂SO₄
MW: 720,75 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD01311715
CAS-nummer: 56391-57-2
EINECS: 260-147-3
Merck Indeks: 14,06479

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Elemental analysis (Nitrogen) 9.20 to 10.20 % (calcd.on dried substance)
Specific rotation [a]20/D +87.0 ~ +96.0 deg (C=3, water, calcd.on dried substance)
Water max. 15.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR