(3-(Naphthalen-1-yl)phenyl)boronic acid (contains varying amounts of Anhydride)
Leverandør: TCI
|
Warning
|
Synonymer:
3-(1-Naphthyl)phenylboronic acid, 3-(1-Naphthyl)benzeneboronic acid
N1131-1G
N1131-5G
TCIAN1131-1GEA
2000
NOK
TCIAN1131-1G
TCIAN1131-5G
(3-(Naphthalen-1-yl)phenyl)boronic acid (contains varying amounts of Anhydride)
(3-(Naphthalen-1-yl)phenyl)boronic acid
Formel:
C₁₆H₁₃BO₂ MW: 248,088 g/mol |
MDL nummer:
MFCD11045054 CAS-nummer: 881913-20-8 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...