1-(2-Nitrophenyl)ethanol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
2-Nitro-alpha-methylbenzyl alcohol
N1132-1G
N1132-5G
TCIAN1132-1GEA
930
NOK
TCIAN1132-1G
TCIAN1132-5G
1-(2-Nitrophenyl)ethanol ≥98.0% (by GC)
1-(2-Nitrophenyl)ethanol
Formel:
C₈H₉NO₃ MW: 167,16 g/mol Kokepunkt: 107°C |
MDL nummer:
MFCD02683380 CAS-nummer: 3205-25-2 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...