N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide ≥98.0% (by HPLC)
Leverandør: TCI
Synonymer:
N-(2-(Cyclohexyloxy)-4-nitrophenyl)methanesulphonamide, NS-398, N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
N1152-10MG
N1152-50MG
TCIAN1152-10MGEA
1575
NOK
TCIAN1152-10MG
TCIAN1152-50MG
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide ≥98.0% (by HPLC)
N-[2-(Cyclohexyloxy)-4-nitrophenyl]methanesulphonamide
Formel:
C₁₃H₁₈N₂O₅S MW: 314,356 g/mol |
MDL nummer:
MFCD00882995 CAS-nummer: 123653-11-2 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 137.0 to 141.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...