Norlidocaine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Monoethylglycinexylidide hydrochloride, N-(2,6-Dimethylphenyl)-2-(ethylamino)acetamide hydrochloride, Norlidocaine HCl

N1171-25MG N1171-100MG
TCIAN1171-25MGEA 3300 NOK
TCIAN1171-25MG TCIAN1171-100MG
Norlidocaine hydrochloride ≥98.0% (by HPLC, titration analysis)
Norlidocaine hydrochloride
Formel: C₁₂H₁₈N₂O·HCl
MW: 242,75 g/mol
MDL nummer: MFCD21334173
CAS-nummer: 7729-94-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR