4-tert-Octylresorcinol ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
4-(1,1,3,3-Tetramethylbutyl)resorcinol, 4-(2,4,4-Trimethylpentan-2-yl)benzene-1,3-diol
O0048-25G
O0048-5G
TCIAO0048-25GEA
15000
NOK
TCIAO0048-25G
TCIAO0048-5G
4-tert-Octylresorcinol ≥99.0% (by GC)
4-tert-Octylresorcinol
Formel:
C₁₄H₂₂O₂ MW: 222,33 g/mol |
MDL nummer:
MFCD00045770 CAS-nummer: 28122-52-3 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 108.0 to 111.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...