(2,7-Octadien-1-yl)succinic anhydride
Leverandør: TCI
|
Warning
|
Synonymer:
3-(Octa-2,7-dien-1-yl)dihydrofuran-2,5-dione, 4,9-Decadiene-1,2-dicarboxylic Anhydride
O0244-25G
O0244-5G
TCIAO0244-25GEA
3300
NOK
TCIAO0244-25G
TCIAO0244-5G
(2,7-Octadien-1-yl)succinic anhydride
(2,7-Octadien-1-yl)succinic anhydride
Formel:
C₁₂H₁₆O₃ MW: 208,26 g/mol |
MDL nummer:
MFCD00191676 CAS-nummer: 142826-45-7 |
Spesifikasjon Testresultater
Purity (Morpholine Method) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...