Octylphosphonic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid

O0380-1G O0380-5G
TCIAO0380-1GEA 570 NOK
TCIAO0380-1G TCIAO0380-5G
Octylphosphonic acid ≥98.0% (by titrimetric analysis)
Octylphosphonic acid
Formel: C₈H₁₉O₃P
MW: 194,21 g/mol
MDL nummer: MFCD00015841
CAS-nummer: 4724-48-5
EINECS: 225-218-5
FN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b975e4d65d03b62396376c96b433a1bc47887d5&vid=4b975e4d65d03b62396376c96b433a1bc47887d5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR