n-Octanal diethyl acetal ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
n-Octyl Aldehyde Diethyl Acetal, 1,1-Diethoxyoctane
O0427-5ML
O0427-25ML
TCIAO0427-5MLEA
340
NOK
TCIAO0427-5ML
TCIAO0427-25ML
n-Octanal diethyl acetal ≥97.0% (by GC)
n-Octanal diethyl acetal
Formel:
C₁₂H₂₆O₂ MW: 202,34 g/mol Kokepunkt: 115°C |
MDL nummer:
MFCD00672807 CAS-nummer: 54889-48-4 EINECS: 259-385-0 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...