n-Octylamine hydrobromide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

O0442-1G O0442-5G
TCIAO0442-1GEA 475 NOK
TCIAO0442-1G TCIAO0442-5G
n-Octylamine hydrobromide ≥98.0% (by titrimetric analysis)
n-Octylamine hydrobromide
Formel: C₈H₁₉N·HBr
Smeltepunkt: 208°C
MDL nummer: MFCD00044802
CAS-nummer: 14846-47-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b4a89e43f077a1e890b50f0c2b56c9ada83fdb4&vid=9b4a89e43f077a1e890b50f0c2b56c9ada83fdb4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR