n-Octylamine hydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: OACl, n-Octylammonium chloride

O0484-1G O0484-5G
TCIAO0484-1GEA 380 NOK
TCIAO0484-1G TCIAO0484-5G
n-Octylamine hydrochloride ≥98.0% (by titrimetric analysis)
n-Octylamine hydrochloride
Formel: C₈H₁₉N·HCl
MW: 165,71 g/mol
Smeltepunkt: 198°C
MDL nummer: MFCD00042019
CAS-nummer: 142-95-0
EINECS: 205-574-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3b499ff2e21ece7929781e0f6c22ed3a66d8f65&vid=c3b499ff2e21ece7929781e0f6c22ed3a66d8f65&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR