Phenolphthalein disulphate tripotassium salt hydrate ≥97.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Phenolphthalein disulphate potassium salt hydrate

P0097-1G P0097-5G
TCIAP0097-1GEA 1850 NOK
TCIAP0097-1G TCIAP0097-5G
Phenolphthalein disulphate tripotassium salt hydrate ≥97.0% (by titrimetric analysis)
Phenolphthalein disulphate tripotassium salt hydrate
Formel: C₂₀H₁₃K₃O₁₁S₂·xH₂O
MW: 610,75 g/mol (anhy)
MDL nummer: MFCD00070538
CAS-nummer: 62625-16-5
EINECS: 263-648-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Water 2.5 to 12.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR