6-Phenyl-2-thiouracil ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 2-Mercapto-6-phenylpyrimidin-4-ol

P0236-1G P0236-5G
TCIAP0236-1GEA 595 NOK
TCIAP0236-1G TCIAP0236-5G
6-Phenyl-2-thiouracil ≥98.0% (by HPLC, titration analysis)
6-Phenyl-2-thiouracil
Formel: C₁₀H₈N₂OS
MW: 204,25 g/mol
MDL nummer: MFCD00059166
CAS-nummer: 36822-11-4
EINECS: 253-227-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR