Phloridzin ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Synonymer:
Phlorizin hydrate
P0248-1G
P0248-5G
TCIAP0248-1GEA
850
NOK
TCIAP0248-1G
TCIAP0248-5G
Phloridzin ≥97.0% (by titrimetric analysis)
Phloridzin
Formel:
C21H24O10 MW: 436,41 g/mol |
MDL nummer:
MFCD00006591 CAS-nummer: 60-81-1 EINECS: 200-487-1 Merck Indeks: 14,07327 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 97.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -52.0 to -55.0 deg (C=3.2, EtOH (95%)) |
Water | max. 8.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...