1-Naphthyl phosphate disodium salt substrate for Phosphatase

Leverandør: TCI
Warning

Synonymer: 1-Naphthylphosphoric acid disodium salt, α-Naphthyl Phosphate Disodium Salt Hydrate, Phosphoric acid 1-Naphthyl ester disodium salt, Disodium 1-Naphthyl phosphate hydrate

P0263-1G P0263-5G
TCIAP0263-1GEA 875 NOK
TCIAP0263-1G TCIAP0263-5G
1-Naphthyl phosphate disodium salt substrate for Phosphatase
1-Naphthyl phosphate disodium salt
Formel: C₁₀H₇Na₂O₄P
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00041007
CAS-nummer: 2183-17-7
EINECS: 218-564-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (Nonaqueous Titration) 97.0 to 103.0 % (calcd.on anh.substance)
Water max. 10.0 %
Alkaline Phosphatase detection test to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR