Triallyl phosphate ≥96.0% (by GC)
Leverandør: TCI
Synonymer:
Phosphoric Acid Triallyl Ester
P0264-25ML
P0264-100ML
TCIAP0264-25MLEA
1300
NOK
TCIAP0264-25ML
TCIAP0264-100ML
Triallyl phosphate ≥96.0% (by GC)
Triallyl phosphate
Formel:
C₉H₁₅O₄P MW: 218,19 g/mol Kokepunkt: 113°C |
MDL nummer:
MFCD00014946 CAS-nummer: 1623-19-4 EINECS: 216-606-5 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...