Piperine ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: (2E,4E)-5-(Benzo[d][1,3]dioxol-5-yl)-1-(piperidin-1-yl)penta-2,4-dien-1-one, 1-[5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoyl]piperidine, 1-Piperoylpiperidine, 1-Piperylpiperidine

P0460-1G P0460-5G
TCIAP0460-1GEA 220 NOK
TCIAP0460-1G TCIAP0460-5G
Piperine ≥97.0% (by HPLC, total nitrogen)
Piperine
Formel: C₁₇H₁₉NO₃
MW: 285,34 g/mol
MDL nummer: MFCD00005839
CAS-nummer: 94-62-2
EINECS: 202-348-0
Merck Indeks: 14,07472

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 128.0 to 133.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR