2-Phenylimidazoline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Phenyl-4,5-dihydro-1H-imidazole, 2-Phenyl-2-imidazoline

P0685-25G P0685-500G
TCIAP0685-25GEA 260 NOK
TCIAP0685-25G TCIAP0685-500G
2-Phenylimidazoline ≥98.0% (by GC, titration analysis)
2-Phenylimidazoline
Formel: C₉H₁₀N₂
MW: 146,19 g/mol
Kokepunkt: 181°C
MDL nummer: MFCD00005180
CAS-nummer: 936-49-2
EINECS: 213-313-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 100.0 to 103.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR