Bis(cis-3,3,5-trimethylcyclohexyl)phthalate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Phthalic Acid Bis(cis-3,3,5-trimethylcyclohexyl) Ester

P0946-25G P0946-5G
TCIAP0946-25GEA 1900 NOK
TCIAP0946-25G TCIAP0946-5G
Bis(cis-3,3,5-trimethylcyclohexyl)phthalate ≥98.0% (by GC)
Bis(cis-3,3,5-trimethylcyclohexyl)phthalate
Formel: C₂₆H₃₈O₄
MW: 414,59 g/mol
MDL nummer: MFCD00070480
CAS-nummer: 245652-81-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 91.0 to 94.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR