Nonanoic anhydride ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Danger
Synonymer: Pelargonic Anhydride
P0957-25ML P0957-500ML
TCIAP0957-25MLEA 1000 NOK
TCIAP0957-25ML TCIAP0957-500ML
Nonanoic anhydride ≥97.0% (by titrimetric analysis)
Nonanoic anhydride
Formel: C₁₈H₃₄O₃
MW: 298,47 g/mol
Kokepunkt: 207°C
MDL nummer: MFCD00057768
CAS-nummer: 1680-36-0
EINECS: 216-854-4
FN: 3265
ADR: 8,III
Bestill nå
Spesifikasjon Testresultater
Appearance Colorless to Light yellow clear liquid
Purity (Morpholine Method) min. 97.0 %