Hexadecyl palmitate ≥97.0% (by GC)
Leverandør: TCI
Synonymer:
Cetyl Palmitate, Palmitic acid palmityl ester, n-Hexadecyl palmitate, Palmitic Acid Hexadecyl Ester, Palmityl palmitate
P1077-25G
P1077-500G
TCIAP1077-25GEA
270
NOK
TCIAP1077-25G
TCIAP1077-500G
Hexadecyl palmitate ≥97.0% (by GC)
Hexadecyl palmitate
Formel:
C₃₂H₆₄O₂ MW: 480,86 g/mol Kokepunkt: 534°C |
MDL nummer:
MFCD00053739 CAS-nummer: 540-10-3 EINECS: 208-736-6 Merck Indeks: 14,02031 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 53.0 to 56.0 deg-C |
Solubility in hot Acetone | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...