Ethyl (phenylthio)acetate ≥97.0% (by GC)
Leverandør: TCI
Synonymer:
(Phenylthio)acetic Acid Ethyl Ester
P1085-25G
P1085-5G
TCIAP1085-25GEA
1550
NOK
TCIAP1085-25G
TCIAP1085-5G
Ethyl (phenylthio)acetate ≥97.0% (by GC)
Ethyl (phenylthio)acetate
Formel:
C₁₀H₁₂O₂S MW: 196,26 g/mol Kokepunkt: 160°C |
MDL nummer:
MFCD00145107 CAS-nummer: 7605-25-6 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...