Trihexyl phosphite ≥95.0% (by GC)
Leverandør: TCI
Synonymer:
Phosphorous Acid Trihexyl Ester
P1260-25G
P1260-5G
TCIAP1260-25GEA
1325
NOK
TCIAP1260-25G
TCIAP1260-5G
Trihexyl phosphite ≥95.0% (by GC)
Trihexyl phosphite
Formel:
C₁₈H₃₉O₃P MW: 334,48 g/mol Kokepunkt: 158°C |
MDL nummer:
MFCD00015282 CAS-nummer: 6095-42-7 EINECS: 228-045-3 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...