D-Proline methyl ester hydrochloride ≥96.0% (by total nitrogen and titration analysis)

Leverandør: TCI
Warning

P1730-1G P1730-5G
TCIAP1730-1GEA 245 NOK
TCIAP1730-1G TCIAP1730-5G
D-Proline methyl ester hydrochloride ≥96.0% (by total nitrogen and titration analysis)
D-Proline methyl ester hydrochloride
Formel: C₆H₁₁NO₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00083685
CAS-nummer: 65365-28-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %
Melting point 70.0 to 76.0 deg-C
Specific rotation [a]20/D +30.0 to +35.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR