5-Phenyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Phenyl-3-(trifluoromethyl)pyrazole

P1756-1G P1756-5G
TCIAP1756-1GEA 540 NOK
TCIAP1756-1G TCIAP1756-5G
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
5-Phenyl-3-(trifluoromethyl)-1H-pyrazole
Formel: C₁₀H₇F₃N₂
MW: 212,18 g/mol
MDL nummer: MFCD00156064
CAS-nummer: 4027-54-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR