Pirfenidone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Methyl-1-phenylpyridin-2(1H)-one, 5-Methyl-1-phenyl-2-pyridone

P1871-1G P1871-100MG
TCIAP1871-1GEA 3800 NOK
TCIAP1871-1G TCIAP1871-100MG
Pirfenidone ≥98.0% (by GC)
Pirfenidone
Formel: C₁₂H₁₁NO
MW: 185,23 g/mol
MDL nummer: MFCD00866047
CAS-nummer: 53179-13-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR