Prostaglandin F2α ≥95.0% (by titrimetric analysis)

Leverandør: TCI
Danger

Synonymer: PGF2alpha, Prostaglandin F2alpha

P1885-1MG P1885-10MG
TCIAP1885-1MGEA 690 NOK
TCIAP1885-1MG TCIAP1885-10MG
Prostaglandin F2α ≥95.0% (by titrimetric analysis)
Prostaglandin F2α
Formel: C₂₀H₃₄O₅
MW: 354,5 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00135231
CAS-nummer: 551-11-1
Merck Indeks: 14,07878

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 95.0 %
Specific rotation [a]20/D +22.0 to +26.0 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR