Pentyl valerate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: Pentanoic Acid Pentyl Ester, Pentyl Pentanoate, Amyl Valerate, Amyl Pentanoate, Valeric Acid Pentyl Ester

P1929-25ML P1929-500ML
TCIAP1929-25MLEA 220 NOK
TCIAP1929-25ML TCIAP1929-500ML
Pentyl valerate ≥98.0% (by GC)
Pentyl valerate
Formel: C₁₀H₂₀O₂
MW: 172,27 g/mol
Kokepunkt: 205°C
Flash punkt: 81°C
MDL nummer: MFCD00042904
CAS-nummer: 2173-56-0
EINECS: 218-528-7

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR