3-Phenylpyrazole-5-carboxylic acid hydrate ≥97.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 5-Phenyl-1H-pyrazole-3-carboxylic acid

P2021-1G P2021-5G
TCIAP2021-1GEA 1500 NOK
TCIAP2021-1G TCIAP2021-5G
3-Phenylpyrazole-5-carboxylic acid hydrate ≥97.0% (by HPLC, titration analysis)
3-Phenylpyrazole-5-carboxylic acid hydrate
Formel: C₁₀H₈N₂O₂
MW: 188,19 g/mol
MDL nummer: MFCD01248821
CAS-nummer: 5071-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0523762b707b51248061d62e4944bca95f2e41d&vid=e0523762b707b51248061d62e4944bca95f2e41d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 % (calcd.on anh.substance)
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR