N-Phthalyl-L-tryptophan ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
(S)-2-(1,3-Dioxoisoindolin-2-yl)-3-(1H-indol-3-yl)propanoic acid, RG 108, RG108
P2023-50MG
P2023-200MG
TCIAP2023-50MGEA
1075
NOK
TCIAP2023-50MG
TCIAP2023-200MG
N-Phthalyl-L-tryptophan ≥98.0% (by HPLC, titration analysis)
N-Phthalyl-L-tryptophan
Formel:
C₁₉H₁₄N₂O₄ MW: 334,33 g/mol Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD08705332 CAS-nummer: 48208-26-0 |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -268 to -273 deg (C=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...