6-Phenyl-3(2H)-pyridazinone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Phenylpyridazin-3(2H)-one, 3-Hydroxy-6-phenylpyridazine
P2060-1G
P2060-5G
TCIAP2060-1GEA
915
NOK
TCIAP2060-1G
TCIAP2060-5G
6-Phenyl-3(2H)-pyridazinone ≥98.0% (by GC)
6-Phenyl-3(2H)-pyridazinone
Formel:
C₁₀H₈N₂O MW: 172,19 g/mol |
MDL nummer:
MFCD00114953 CAS-nummer: 2166-31-6 EINECS: 218-505-1 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 201.0 to 205.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...