Psoralen ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 7H-Furo[3,2-g]chromen-7-one, Ficusin

P2077-20MG P2077-100MG
TCIAP2077-20MGEA 915 NOK
TCIAP2077-20MG TCIAP2077-100MG
Psoralen ≥99.0% (by GC)
Psoralen
Formel: C₁₁H₆O₃
MW: 186,17 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00010520
CAS-nummer: 66-97-7
EINECS: 200-639-7
Merck Indeks: 14,07928
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66cecac710c498d0b23dec0442d0c7e096aef1ba&vid=66cecac710c498d0b23dec0442d0c7e096aef1ba&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 99.0 %
Melting point 162.0 to 166.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR