Pentafluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI
Synonymer:
(S)-2-Amino-3-(perfluorophenyl)propanoic acid
P2085-200MG
P2085-1G
TCIAP2085-200MGEA
700
NOK
TCIAP2085-200MG
TCIAP2085-1G
Pentafluoro-L-phenylalanine ≥95.0% (by HPLC, titration analysis)
Pentafluoro-L-phenylalanine
Formel:
C₉H₆F₅NO₂ MW: 255,14 g/mol |
MDL nummer:
MFCD01860881 CAS-nummer: 34702-59-5 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Specific rotation [a]20/D | +18.0 to +23.0 deg (C=0.2,H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...