2-(1H-Pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3-(2-Hydroxyphenyl)pyrazole

P2218-1G P2218-5G
TCIAP2218-1GEA 595 NOK
TCIAP2218-1G TCIAP2218-5G
2-(1H-Pyrazol-3-yl)phenol ≥98.0% (by GC, titration analysis)
2-(1H-Pyrazol-3-yl)phenol
Formel: C₉H₈N₂O
MW: 160,18 g/mol
MDL nummer: MFCD03093876
CAS-nummer: 34810-67-8
EINECS: 252-222-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28eb6abe6bc581cae8a326a720a7d50b93401d6a&vid=1dc7b7f682b131c5002a6b53eaf84c6eefe81e14&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR