4-(2-Propynyloxy)aniline ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Aminophenyl Propargyl ether, 4-(Propargyloxy)aniline

P2224-1G P2224-5G
TCIAP2224-1GEA 1700 NOK
TCIAP2224-1G TCIAP2224-5G
4-(2-Propynyloxy)aniline ≥98.0% (by GC, titration analysis)
4-(2-Propynyloxy)aniline
Formel: C₉H₉NO
MW: 147,18 g/mol
Kokepunkt: 95°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD12823190
CAS-nummer: 26557-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c4465c3041aafef9146101fcd83394a2769b033&vid=1c4465c3041aafef9146101fcd83394a2769b033&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR