1-(2-Propynyl)piperazine ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 1-(Prop-2-yn-1-yl)piperazine, 1-Propargylpiperazine

P2228-200MG P2228-1G
TCIAP2228-200MGEA 1200 NOK
TCIAP2228-200MG TCIAP2228-1G
1-(2-Propynyl)piperazine ≥97.0% (by GC, titration analysis)
1-(2-Propynyl)piperazine
Formel: C₇H₁₂N₂
MW: 124,19 g/mol
Kokepunkt: 90°C
MDL nummer: MFCD08445213
CAS-nummer: 52070-67-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=930b68c94f33ce38066b345cce6d8976aae54105&vid=930b68c94f33ce38066b345cce6d8976aae54105&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 58.0 to 62.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR