Pentafluorophenyl Benzoate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Benzoic acid Pentafluorophenyl ester

P2229-1G P2229-5G
TCIAP2229-1GEA 525 NOK
TCIAP2229-1G TCIAP2229-5G
Pentafluorophenyl Benzoate ≥98.0% (by GC)
Pentafluorophenyl Benzoate
Formel: C₁₃H₅F₅O₂
MW: 288,17 g/mol
CAS-nummer: 1548-84-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40e3c2901bbb936acd61c68ec1dc2889f1f59c11&vid=40e3c2901bbb936acd61c68ec1dc2889f1f59c11&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR